Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Accessory Protein (IL1RAP) CLIA Kit
Supplier:  Biomatik Corporation EKU09650
Encompass_Preferred
Mouse Interleukin 1 Receptor Accessory Protein (IL1RAP) CLIA Kit for detecting IL1RAP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.95pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q61730. Gene ID: 16180. Target Synonyms: IL-1RAcP; C3orf13; IL1R3; Interleukin-1 receptor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5732