Learn More
American Research Products Inc Mouse Interleukin 1 receptor l
Catalog No. 5000327347
Encompass
Mouse Interleukin-1 receptor-like 1 (IL1RL1) ELISA kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27347 Supplier American Research Products Inc Supplier No. CSBEL011626MO
Mouse Interleukin-1 receptor-like 1 (IL1RL1) ELISA kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich