Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Type I (IL1R1) ELISA Kit
Supplier:  Biomatik Corporation EKU05167
Encompass_Preferred
Mouse Interleukin 1 Receptor Type I (IL1R1) ELISA Kit for detecting IL1R1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 14.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13504. Gene ID: 16177. Target Synonyms: CD121a; CD121-A; IL1-R1; IL1R; IL1RA; P80; CD121 Antigen-Like Family Member A; Interleukin-1 Receptor Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5501