Learn More
Cusabio Technology LLC Mouse interleukin 10IL10 ELISA
Catalog No. 5000049810
Encompass_Preferred
This kit detects IL10 in mouse serum plasma and tissue homogenates with a sensitivity of 0 78 pg/mL and a quantitative detection range of 3 12 pg/mL to 200 pg/mL Utilizing a sandwich ELISA methodology it provides precise measurements within 1-5 hours using 50-100 L of sample per test The assay employs specific antibodies to capture and detect IL10 followed by colorimetric analysis at 450 nm for accurate readouts Designed for flexibility it accommodates diverse biological matrices while maintaining high reproducibility making it suitable for research applications requiring reliable interleukin-10 quantification in murine models The standardized protocol ensures consistent performance across experimental setups minimizing variability in results
Catalog No. 50-000-49810 Supplier Cusabio Technology LLC Supplier No. CSBE04594MIS
This kit detects IL10 in mouse serum plasma and tissue homogenates with a sensitivity of 0 78 pg/mL and a quantitative detection range of 3 12 pg/mL to 200 pg/mL Utilizing a sandwich ELISA methodology it provides precise measurements within 1-5 hours using 50-100 L of sample per test The assay employs specific antibodies to capture and detect IL10 followed by colorimetric analysis at 450 nm for accurate readouts Designed for flexibility it accommodates diverse biological matrices while maintaining high reproducibility making it suitable for research applications requiring reliable interleukin-10 quantification in murine models The standardized protocol ensures consistent performance across experimental setups minimizing variability in results