Learn More
Biomatik Corporation Mouse Interleukin 11 (IL11) CLIA Kit
Supplier:  Biomatik Corporation EKU08792
Encompass_Preferred
Mouse Interleukin 11 (IL11) CLIA Kit for detecting IL11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P47873. Gene ID: 16156. Target Synonyms: AGIF; Adipogenesis Inhibitory Factor; Oprelvekin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5603