Learn More
American Research Products Inc Mouse interleukin 12 IL 12 P4
Catalog No. 5000326278
Encompass
Mouse interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26278 Supplier American Research Products Inc Supplier No. CSBE07360M
Mouse interleukin 12 IL-12/P40 ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich