Learn More
American Research Products Inc Mouse Interleukin 12 IL 12 P7
Catalog No. 5000326090
Encompass
Mouse Interleukin 12 IL-12/P70 ELISA KIT from CUSABIO Detection Range 62 5 pg/ml - 4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26090 Supplier American Research Products Inc Supplier No. CSBE04600M
Mouse Interleukin 12 IL-12/P70 ELISA KIT from CUSABIO Detection Range 62 5 pg/ml - 4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich