Learn More
American Research Products Inc Mouse Interleukin 12 IL 12 P7
Supplier:  American Research Products Inc CSBE04600MIS
Encompass
Mouse Interleukin 12 IL-12/P70 ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26384