Learn More
Biomatik Corporation Mouse Interleukin 13 Receptor Alpha 2 (IL13Ra2) ELISA Kit
Supplier:  Biomatik Corporation EKU05224
Encompass_Preferred
Mouse Interleukin 13 Receptor Alpha 2 (IL13Ra2) ELISA Kit for detecting IL13Ra2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88786. Gene ID: 16165. Target Synonyms: CD213A2; CD213-A2; IL13R-A2; IL13-RA2; IL13-R; IL13-BP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8911