Learn More
Biomatik Corporation Mouse Interleukin 16 (IL16) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11171
Encompass_Preferred
Mouse Interleukin 16 (IL16) Mini sample ELISA Kit for detecting IL16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54824. Gene ID: 16170. Target Synonyms: LCF; PrIL-16; Lymphocyte Chemoattractant Factor; Pro-Interleukin 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6642