Learn More
American Research Products Inc Mouse Interleukin 17 IL 17 EL
Catalog No. 5000326299
Encompass
Mouse Interleukin 17 IL-17 ELISA Kit from CUSABIO Detection Range 10 938 pg/mL-700 pg/mL Sensitivity 5 469 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26299 Supplier American Research Products Inc Supplier No. CSBE04608MMS
Mouse Interleukin 17 IL-17 ELISA Kit from CUSABIO Detection Range 10 938 pg/mL-700 pg/mL Sensitivity 5 469 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich