Learn More
American Research Products Inc Mouse Interleukin 17F IL17F
Catalog No. 5000327668
Encompass
Mouse Interleukin-17F (IL17F) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27668 Supplier American Research Products Inc Supplier No. CSBEL011601MO
Mouse Interleukin-17F (IL17F) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich