Learn More
American Research Products Inc Mouse interleukin 18 binding p
Catalog No. 5000328550
Encompass
Mouse interleukin-18 binding prorein (IL-18BP) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-28550 Supplier American Research Products Inc Supplier No. CSBE17797M
Mouse interleukin-18 binding prorein (IL-18BP) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich