Learn More
American Research Products Inc Mouse Interleukin 1b IL 1b E
Catalog No. 5000326125
Encompass
Mouse Interleukin 1b (IL-1b) ELISA Kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26125 Supplier American Research Products Inc Supplier No. CSBE08054M1
Mouse Interleukin 1b (IL-1b) ELISA Kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich