Learn More
Biomatik Corporation Mouse Interleukin 2 (IL2) CLIA Kit
Supplier:  Biomatik Corporation EKU08798
Encompass_Preferred
Mouse Interleukin 2 (IL2) CLIA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.14pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04351. Gene ID: 16183. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6963