Learn More
Bioworld MOUSE INTERLEUKIN-2 (RECOMB
Supplier:  Bioworld 220605941
Encompass_Preferred
MOUSE INTERLEUKIN-2 (RECOMBINANT): Recombinant Murine Interleukin-2 Source: E.Coli Description: Recombinant Murine IL-2 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 149 amino acids and having a molecu...
Catalog No. 50-253-694