Learn More
American Research Products Inc Mouse Interleukin 20 IL20 EL
Catalog No. 5000326887
Encompass
Mouse Interleukin-20 (IL20) ELISA kit from CUSABIO Detection Range 31 25 pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26887 Supplier American Research Products Inc Supplier No. CSBEL011630MO
Mouse Interleukin-20 (IL20) ELISA kit from CUSABIO Detection Range 31 25 pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich