Learn More
American Research Products Inc Mouse Interleukin 21 IL 21 E
Catalog No. 5000329868
Encompass
Mouse Interleukin 21 (IL-21) ELISA Kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29868 Supplier American Research Products Inc Supplier No. CSBE13162M
Mouse Interleukin 21 (IL-21) ELISA Kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich