Learn More
American Research Products Inc Mouse Interleukin 22 IL 22 E
Supplier:  American Research Products Inc CSBE13513MIS
Encompass
Mouse Interleukin 22 (IL-22) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-29397