Learn More
Biomatik Corporation Mouse Interleukin 22 (IL22) CLIA Kit
Supplier:  Biomatik Corporation EKU09885
Encompass_Preferred
Mouse Interleukin 22 (IL22) CLIA Kit for detecting IL22 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9JJY9. Gene ID: 50929. Target Synonyms: ILTIF; Zcyto18; IL-TIF; IL-D110; TIFa; TIFIL-23; IL-22; Cytokine Zcyto18; IL-10-Related T-Cell-Derived Inducible Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4842