Learn More
American Research Products Inc Mouse Interleukin 22 receptor
Catalog No. 5000328322
Encompass
Mouse Interleukin-22 receptor subunit alpha-2 (IL22RA2) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28322 Supplier American Research Products Inc Supplier No. CSBEL011637MO
Mouse Interleukin-22 receptor subunit alpha-2 (IL22RA2) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich