Learn More
American Research Products Inc Mouse Interleukin 23 IL 23 EL
Catalog No. 5000329446
Encompass
Mouse Interleukin 23 IL-23 ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29446 Supplier American Research Products Inc Supplier No. CSBE08463M
Mouse Interleukin 23 IL-23 ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich