Learn More
Biomatik Corporation Mouse Interleukin 27A (IL27A) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11189
Encompass_Preferred
Mouse Interleukin 27A (IL27A) Mini sample ELISA Kit for detecting IL27A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K3I6. Gene ID: 246779. Target Synonyms: p28; IL30; Interleukin 30; Interleukin-27 subunit alpha; IL-27 subunit alpha; IL27-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8200