Learn More
Biomatik Corporation Mouse Interleukin 34 (IL34) ELISA Kit
Supplier:  Biomatik Corporation EKU05325
Encompass_Preferred
Mouse Interleukin 34 (IL34) ELISA Kit for detecting IL34 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R1R4. Gene ID: 76527. Target Synonyms: C16orf77. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5914