Learn More
American Research Products Inc Mouse Interleukin 35 IL 35 E
Supplier:  American Research Products Inc CSBE13145M
Encompass
Mouse Interleukin 35 (IL-35) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29049