Learn More
Biomatik Corporation Mouse Interleukin 35 (IL35) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11198
Encompass_Preferred
Mouse Interleukin 35 (IL35) Mini sample ELISA Kit for detecting IL35 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43431. Gene ID: 16159. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5756