Learn More
American Research Products Inc Mouse Interleukin 36 beta IL1
Supplier:  American Research Products Inc CSBEL011619MO
Encompass
Mouse Interleukin-36 beta (IL1F8) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27330