Learn More
Biomatik Corporation Mouse Interleukin 4 (IL4) CLIA Kit
Supplier:  Biomatik Corporation EKU08801
Encompass_Preferred
Mouse Interleukin 4 (IL4) CLIA Kit for detecting IL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P07750. Gene ID: 16189. Target Synonyms: BSF1; BCGF1 ;B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8628