Learn More
Bioworld MOUSE INTERLEUKIN-4 (RECOMB
Supplier:  Bioworld 220605961
Encompass_Preferred
MOUSE INTERLEUKIN-4 (RECOMBINANT): Recombinant Murine Interleukin-4 (IL-4) Source: E.Coli Description: Recombinant Mouse IL-4 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 120 amino acids and having a ...
Catalog No. 50-253-696