Learn More
Biomatik Corporation Mouse Interleukin 5 Receptor Alpha (IL5Ra) ELISA Kit
Supplier:  Biomatik Corporation EKU05353
Encompass_Preferred
Mouse Interleukin 5 Receptor Alpha (IL5Ra) ELISA Kit for detecting IL5Ra in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.50-4000pg/ml. Sensitivity: 28.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21183. Gene ID: 16192. Target Synonyms: CDw125; CD125; IL5R-A; IL5-RA; HSIL5R3; IL5R; IL5R-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8910