Learn More
American Research Products Inc Mouse Interleukin 6 IL 6 ELI
Catalog No. 5000326368
Encompass
Mouse Interleukin 6 (IL-6) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26368 Supplier American Research Products Inc Supplier No. CSBE04639M1
Mouse Interleukin 6 (IL-6) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich