Learn More
American Research Products Inc Mouse Interleukin 7 receptor s
Catalog No. 5000328089
Encompass
Mouse Interleukin-7 receptor subunit alpha (IL7R) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28089 Supplier American Research Products Inc Supplier No. CSBEL011670MO
Mouse Interleukin-7 receptor subunit alpha (IL7R) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich