Learn More
Raybiotech Inc G-Series Mouse Interleukin Array 1000 Kit.
Catalog No. 501950467
Encompass_Preferred
A combination of 2 non-overlapping arrays to measure the relative expression levels of 30 mouse interleukins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-17A, IL-17B, IL-17E (IL-25), IL-17F, IL-2, IL-2 R alpha, IL-20, IL-21, IL-23 p19, IL-28A (IFN-lambda 2), IL-3, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-9, TNF alpha
Catalog No. 50-195-0467 Supplier Raybiotech Inc Supplier No. GSMINT10002
A combination of 2 non-overlapping arrays to measure the relative expression levels of 30 mouse interleukins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-17A, IL-17B, IL-17E (IL-25), IL-17F, IL-2, IL-2 R alpha, IL-20, IL-21, IL-23 p19, IL-28A (IFN-lambda 2), IL-3, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-9, TNF alpha