Learn More
Raybiotech Inc Quantibody Mouse Interleukin Array 1000 Kit.
Catalog No. 501950888
Encompass_Preferred
A combination of Mouse Interleukin Array Q1 and Mouse Interleukin Array Q2. Detects 30 Mouse Interleukins. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-17A, IL-17B, IL-17E (IL-25), IL-17F, IL-2, IL-2 R alpha, IL-20, IL-21, IL-23 p19, IL-28A (IFN-lambda 2), IL-3, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-9, TNF alpha
Catalog No. 50-195-0888 Supplier Raybiotech Inc Supplier No. QAMINT10002
A combination of Mouse Interleukin Array Q1 and Mouse Interleukin Array Q2. Detects 30 Mouse Interleukins. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-17A, IL-17B, IL-17E (IL-25), IL-17F, IL-2, IL-2 R alpha, IL-20, IL-21, IL-23 p19, IL-28A (IFN-lambda 2), IL-3, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-9, TNF alpha