Learn More
Raybiotech Inc Quantibody Mouse Interleukin Array 2 Kit.
Supplier:  Raybiotech Inc QAMINT22
Encompass_Preferred
Detects 10 Mouse Interleukins. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IL-1 ra (IL-1 F3), IL-2 R alpha, IL-6 R, IL-11, IL-12 p40, IL-17B, IL-17E (IL-25), IL-17F, IL-20, IL-28A (IFN-lambda 2)
Catalog No. 50-195-0892