Learn More
Alzet ALZETTM Mouse Intrathecal Catheter: Optimized for Intrathecal Delivery from any ALZET Osmotic Pump
SDP
Catalog No. NC0626848
Encompass
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous delivery of test agents to the mouse intrathecal space. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 6 cm total length; distal tip 0.75 Fr; internal volume ~8.6 μlOptimum Animal Size: Adult mouseFeatures & Benefits:•ALZET connection can be trimmed•Includes a Teflon-coated stylet for easier placement•ALZET connection provides secure fit to any ALZET pump
Catalog No. NC0626848 Supplier Alzet Supplier No. 0007743
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous delivery of test agents to the mouse intrathecal space. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 6 cm total length; distal tip 0.75 Fr; internal volume ~8.6 μlOptimum Animal Size: Adult mouseFeatures & Benefits:•ALZET connection can be trimmed•Includes a Teflon-coated stylet for easier placement•ALZET connection provides secure fit to any ALZET pump