Learn More
Biomatik Corporation Mouse Ionized Calcium-binding Adapter Molecule 1 (IBA1) CLIA Kit
Supplier:  Biomatik Corporation EKU09826
Encompass_Preferred
Mouse Ionized Calcium-binding Adapter Molecule 1 (IBA1) CLIA Kit for detecting IBA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O70200. Gene ID: 11629. Target Synonyms: AIF1; IRT1; Daintain; Protein G1; Allograft Inflammatory Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5964