Learn More
FUJIFILM BIOSCIENCES INC Mouse IP-10 (CXCL10), Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200381MG
Encompass_Preferred
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells, and fibroblasts in response to interferon gamma (IFN-Y).  IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3.  IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus. 
Catalog No. 50-990-868