Learn More
BIOHIPPO Mouse Irisin Microsample
SDP
Catalog No. 5000314926
Encompass
Mouse Irisin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Irisin in Serum plasma and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 18 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Irisin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Irisin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Irisin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14926 Supplier BIOHIPPO Supplier No. BHE15212480
Add to Cart
Add to Cart
Mouse Irisin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Irisin in Serum plasma and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 18 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Irisin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Irisin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Irisin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color