Learn More
Braintree Scientific Mouse Irradiation Chamber
SDP
Catalog No. 501954180
Encompass
Made from (3/16") clear acrylic, the mouse irradiation chamber features a flip-top lid for easy loading and unloading. Perforated top allows air to circulate throughout the unit. Top incorporates a slot for filter material to improve contamination control. Dimensions: 6" Dia. x 2" H **Sold for research purposes only and not for human use**
Catalog No. 50-195-4180 Supplier Braintree Scientific Supplier No. AKMIC
Made from (3/16") clear acrylic, the mouse irradiation chamber features a flip-top lid for easy loading and unloading. Perforated top allows air to circulate throughout the unit. Top incorporates a slot for filter material to improve contamination control. Dimensions: 6" Dia. x 2" H **Sold for research purposes only and not for human use**