Learn More
Crystal Chem Inc Mouse Monoclonal Isotyping Kit w/Kappa & Lambda,10 tests
SDP
Catalog No. 501947759
Encompass
The Mouse RapidYield Monoclonal Antibody Isotyping Kit is a highly sensitive, 10 minute rapid lateral flow isotyping kit for determining mouse monoclonal antibody classes.Highlights: Five minute, rapid lateral flow isotyping kit. High sensitivity of 10 ng/mL. For cell/tissue culture supernatant, mouse ascites fluid, or purified antibodies. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7759 Supplier Crystal Chem Inc Supplier No. 80601
The Mouse RapidYield Monoclonal Antibody Isotyping Kit is a highly sensitive, 10 minute rapid lateral flow isotyping kit for determining mouse monoclonal antibody classes.Highlights: Five minute, rapid lateral flow isotyping kit. High sensitivity of 10 ng/mL. For cell/tissue culture supernatant, mouse ascites fluid, or purified antibodies. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.