Learn More
Bon Opus Biosciences Recombinant Mouse Jagged1 Protein (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP104650UG
Encompass_Preferred
Recombinant Mouse Jagged1 Protein (C-hFc) is produced by Human Cells. The target gene encoding Gln34-Ser1046 is expressed with a C-hFc tag. Accession# P78504-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1046
Catalog No. 50-217-2645