Learn More
SAI Infusion Technologies Mouse Jugular Vein Catheter - for mice <25g
SDP
Catalog No. NC0133052
Encompass
Jugular Vein Catheter for Mice under 25g. Polyurethane, 9mm 1Fr tip, 2 suture beads, 14cm total length. Access with 25G blunt needle (such as B25-50). Sterile. Catheters may be used for sampling or infusion. Our biocompatible polyurethane catheters are designed to enhance patency and minimize vessel trauma during implantation. Retention (suture) beads are accurately positioned from the catheter tip for optimal implantation position within the vessel. Beads are also positioned to provide a secure tension loop to maximize continued patency during movement and growth. Product is for laboratory animal research only.
Catalog No. NC0133052 Supplier SAI Infusion Technologies Supplier No. MJC01
 Restricted Item
Add to Cart
Add to Cart
Jugular Vein Catheter for Mice under 25g. Polyurethane, 9mm 1Fr tip, 2 suture beads, 14cm total length. Access with 25G blunt needle (such as B25-50). Sterile. Catheters may be used for sampling or infusion. Our biocompatible polyurethane catheters are designed to enhance patency and minimize vessel trauma during implantation. Retention (suture) beads are accurately positioned from the catheter tip for optimal implantation position within the vessel. Beads are also positioned to provide a secure tension loop to maximize continued patency during movement and growth. Product is for laboratory animal research only.