Learn More
BIOHIPPO Mouse JUNB Microsample
SDP
Catalog No. 5000315445
Encompass
Mouse JUNB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of JUNB in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 074 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse JUNB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse JUNB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse JUNB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15445 Supplier BIOHIPPO Supplier No. BHE15211208
Mouse JUNB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of JUNB in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 074 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse JUNB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse JUNB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse JUNB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color