Learn More
Biomatik Corporation Mouse Kallikrein 13 (KLK13) ELISA Kit
Supplier:  Biomatik Corporation EKU05434
Encompass_Preferred
Mouse Kallikrein 13 (KLK13) ELISA Kit for detecting KLK13 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36368. Gene ID: 13647. Target Synonyms: KLK-L4; KLKL4; Kallikrein-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6858