Learn More
Sino Biological Recombinant Anti-VEGFR2/KDR Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50998R001P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse VEGFR2 / Flk-1 / CD309 / KDR (rM VEGFR2 / Flk-1 / CD309 / KDR; Catalog#50998-M08H; P35918-1; Met1-Glu762) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8788