Learn More
Cusabio Technology LLC Mouse Keratin, type I cytoskeletal 18 fragment (K18-Asp396)ELISA Kit
Supplier:  Cusabio Technology LLC CSBE14265M
Encompass_Preferred
This Mouse CK 18-M30 ELISA Kit was designed for the quantitative measurement of Mouse CK 18-M30 protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 15.6 μIU/mL-1000 μIU/mL and the sensitivity is 3.9 μIU/mL. One kit contains 96 wells.
Catalog No. NC2862348