Learn More
Cusabio Technology LLC Mouse KERATIN KRT18 ELISA Kit, 96T, Quantitative Sandwich ELISA, Serum/Plasma, Detection Range 0.156 ng/mL - 10 ng/mL, Sensitivity 0.039 ng/mL
Supplier:  Cusabio Technology LLC CSBE17158M
Encompass_Preferred
Mouse KERATIN KRT18 ELISA Kit, 96T, Quantitative Sandwich ELISA, Serum/Plasma, Detection Range 0.156 ng/mL - 10 ng/mL, Sensitivity 0.039 ng/mL
Catalog No. NC2862349