Learn More
American Research Products Inc Mouse Keratinocyte Growth Fact
Catalog No. 5000329297
Encompass
Mouse Keratinocyte Growth Factor (KGF/FGF-7) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29297 Supplier American Research Products Inc Supplier No. CSBE13046M
Mouse Keratinocyte Growth Factor (KGF/FGF-7) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich