Learn More
American Research Products Inc Mouse Kidney injury molecule 1
Catalog No. 5000326212
Encompass
Mouse Kidney injury molecule 1 Kim-1 ELISA Kit from CUSABIO Detection Range 0 78 pg/ml-50 pg/ml Sensitivity 0 195 pg/ml Sample serum plasma urine tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-26212 Supplier American Research Products Inc Supplier No. CSBE08809MIS
Mouse Kidney injury molecule 1 Kim-1 ELISA Kit from CUSABIO Detection Range 0 78 pg/ml-50 pg/ml Sensitivity 0 195 pg/ml Sample serum plasma urine tissue homogenates cell culture supernates Method Sandwich